Learn More
Cayman Chemical Dorsomorphin (hydrochloride), Purity: ≥98%, CAS: 1219168-18-9, 10 mg
SDP
Catalog No. 502985499
Encompass_Preferred
A potent, reversible inhibitor of AMPK (Ki = 109 nM); dose-dependently inhibits the bone morphogenetic protein type 1 receptors ACTR-I (ALK2), BMPR-IA (ALK3), and BMPR-IB (ALK6); downregulates the Akt/mTOR pathway to induce autophagy in U251 human glioma cells
Catalog No. 50-298-5499 Supplier Cayman Chemical Supplier No. 2120710 MG
A potent, reversible inhibitor of AMPK (Ki = 109 nM); dose-dependently inhibits the bone morphogenetic protein type 1 receptors ACTR-I (ALK2), BMPR-IA (ALK3), and BMPR-IB (ALK6); downregulates the Akt/mTOR pathway to induce autophagy in U251 human glioma cells