Learn More
Medchemexpress LLC HY-109030 10mg Medchemexpress, Dorzagliatin CAS:1191995-00-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10903010MG
Encompass_Preferred
Medchemexpress, HY-109030 10mg Dorzagliatin CAS:1191995-00-2 Dorzagliatin (HMS5552), a dual-acting glucokinase (GK) activator, improves glycaemic control and pancreatic β-cell function in type 2 diabetes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9656