Learn More
Apexbio Technology LLC Dorzolamide HCl 130693-82-2 10mM (in 1mL H2O)
SDP
Catalog No. 5000093757
Encompass_Preferred
Dorzolamide hydrochloride (CAS 130693-82-2) is a synthetic small molecule that selectively inhibits carbonic anhydrase an enzyme involved in the reversible hydration of carbon dioxide By blocking this enzyme s activity Dorzolamide HCl reduces the production of aqueous humor which influences intraocular pressure This mechanism underpins its utility in studies investigating ocular physiology glaucoma pathogenesis and the pharmacological modulation of carbonic anhydrase Dorzolamide HCl serves as a valuable tool for elucidating carbonic anhydrase-related pathways in preclinical and biochemical research contexts
Catalog No. 50-000-93757 Supplier Apexbio Technology LLC Supplier No. A43575.1
Dorzolamide hydrochloride (CAS 130693-82-2) is a synthetic small molecule that selectively inhibits carbonic anhydrase an enzyme involved in the reversible hydration of carbon dioxide By blocking this enzyme s activity Dorzolamide HCl reduces the production of aqueous humor which influences intraocular pressure This mechanism underpins its utility in studies investigating ocular physiology glaucoma pathogenesis and the pharmacological modulation of carbonic anhydrase Dorzolamide HCl serves as a valuable tool for elucidating carbonic anhydrase-related pathways in preclinical and biochemical research contexts