Learn More
Selleck Chemical LLC Dorzolamide HCl 10mg 130693-82-2 MK-507 (L-671152) HCl
SDP
Catalog No. 507025
Encompass_Preferred
Dorzolamide (MK-507, L-671152) HCl is a water-soluble, potent inhibitor of human carbonic anhydrase II and IV with Ki of 1.9 nM and 31 nM, respectively, used as anti-glaucoma agent. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-702-5 Supplier Selleck Chemical LLC Supplier No. S137510MG
Dorzolamide (MK-507, L-671152) HCl is a water-soluble, potent inhibitor of human carbonic anhydrase II and IV with Ki of 1.9 nM and 31 nM, respectively, used as anti-glaucoma agent. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.