Learn More
Medchemexpress LLC Dorzolamide (hydrochloride) | 130693-82-2 | 99.9% | C10H17ClN2O4S3 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0109A
Encompass_Preferred
Dorzolamide hydrochloride is a potent carbonic anhydrase II inhibitor, with IC50 values of 0.18 nM and 600 nM for red blood cell CA-II and CA-I respectively. It also exhibits anti-tumor activity.
  • Potent carbonic anhydrase II inhibitor.
  • Possesses anti-tumor activity.
  • Effective in retarding the growth of EAC in mice.
  • Upregulates TXNIP and p53 while downregulating bcl-2.
Catalog No. 50-003-58706