Learn More
Medchemexpress LLC Dorzolamide hydrochloride | 130693-82-2 | MFCD00884659 | 99.9% | C10H17ClN2O4S3 | 5 MG
SDP
Catalog No. 5000433524
Encompass_Preferred
Dorzolamide (L671152) hydrochloride is a potent carbonic anhydrase II inhibitor. It effectively inhibits red blood cell CA-II with an IC50 value of 0.18 nM and CA-I with an IC50 of 600 nM. This compound also exhibits anti-tumor activity.
  • Potent carbonic anhydrase II inhibitor
  • IC50 values of 0.18 nM for red blood cell CA-II
  • IC50 values of 600 nM for CA-I
  • Exhibits anti-tumor activity
Catalog No. 50-004-33524 Supplier Medchemexpress LLC Supplier No. HYB0109A5MG
Dorzolamide (L671152) hydrochloride is a potent carbonic anhydrase II inhibitor. It effectively inhibits red blood cell CA-II with an IC50 value of 0.18 nM and CA-I with an IC50 of 600 nM. This compound also exhibits anti-tumor activity.
  • Potent carbonic anhydrase II inhibitor
  • IC50 values of 0.18 nM for red blood cell CA-II
  • IC50 values of 600 nM for CA-I
  • Exhibits anti-tumor activity