Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Dose Makes Poison: Testing Environmental Impacts of Pollution Experiment Kit
SDPSCI_ED

Catalog No. S42876 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S42876 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S42876 Supplier Edvotek™ Supplier No. 905
Add to Cart
Add to Cart
This item is not returnable. View return policy

Allows scientists to determine the effects of given substance on living organisms.

  • Biological assay for environmental pollution
  • Inquiry-based lab students plan and perform plant bioassay
  • Determines environmental hazard of common point and non-point source pollutants
  • Results analyzed using averages, standard deviations, TC50 calculations, integrating STEM

Specifications

Grade 9 to Undergraduate
For Use With (Application) Educational purpose
Includes Instructions, radish seeds, plant growth medium, antifungal powder, pollutant solutions, cheese cloth, petri dishes, transfer pipets, serological pipet, conical tubes, microcentrifuge tubes, small loops
Time Range 2 hours, 15 minutes (plus one week for growing)
Content And Storage Refrigeration and freezer storage
Class Size 10 Lab Groups
Product Type Environmental Pollution Bioassay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.