Learn More
Cayman Chemical DOSEHchlorIde 10mg
SDP
Catalog No. 502978421
Encompass_Preferred
A cationic sulfonium lipid; has been used in the generation of sLNPs for the delivery of mRNA in vivo; LNPs containing DOSEH and encapsulating an mRNA reporter accumulate primarily in the lungs after intravenous administration in mice; DOSEH-containing LNPs encapsulating Cre mRNA induce the expression of tdTomato in the lungs of Ai14 mice engineered to express tdTomato upon Cre-mediated recombination
Catalog No. 50-297-8421 Supplier Cayman Chemical Supplier No. 4160110 MG
A cationic sulfonium lipid; has been used in the generation of sLNPs for the delivery of mRNA in vivo; LNPs containing DOSEH and encapsulating an mRNA reporter accumulate primarily in the lungs after intravenous administration in mice; DOSEH-containing LNPs encapsulating Cre mRNA induce the expression of tdTomato in the lungs of Ai14 mice engineered to express tdTomato upon Cre-mediated recombination