Learn More
Medchemexpress LLC Dosimertinib-d5 mesylate | 2403760-72-3 | 99.4% | 600.74 g·mol⁻1 | C29H32D5N7O5S | 5 MG
SDP
Catalog No. 5000220979
Encompass_Preferred
Dosimertinib-d5 mesylate is the deuterated mesylate salt of an EGFR inhibitor intended for preclinical research. It is reported to reduce phosphorylation of EGFR and ERK, show antiproliferative and antitumor activity in model systems, and is used in studies of non-small-cell lung cancer and EGFR signaling pathways.
  • Deuterated small molecule designed for EGFR pathway research.
  • Mesylate salt form with improved handling and solubility characteristics.
  • Demonstrated decrease in p-EGFR and p-ERK levels in vitro.
  • Shows antiproliferative and antitumor activity in preclinical models.
  • Supplied in a 5 mg package suitable for analytical and in vitro studies.
Catalog No. 50-002-20979 Supplier Medchemexpress LLC Supplier No. HY142283AS5MG
Dosimertinib-d5 mesylate is the deuterated mesylate salt of an EGFR inhibitor intended for preclinical research. It is reported to reduce phosphorylation of EGFR and ERK, show antiproliferative and antitumor activity in model systems, and is used in studies of non-small-cell lung cancer and EGFR signaling pathways.
  • Deuterated small molecule designed for EGFR pathway research.
  • Mesylate salt form with improved handling and solubility characteristics.
  • Demonstrated decrease in p-EGFR and p-ERK levels in vitro.
  • Shows antiproliferative and antitumor activity in preclinical models.
  • Supplied in a 5 mg package suitable for analytical and in vitro studies.