Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dosimertinib-d5 mesylate | 2403760-72-3 | 99.4% | C29H32D5N7O5S | 10MG
SDP

Catalog No. 5000261997
Encompass_Preferred

Dosimertinib-d5 mesylate is the deuterated mesylate salt of dosimertinib, a third-generation epidermal growth factor receptor (EGFR) inhibitor. Provided as a research-grade reference standard for preclinical and analytical applications; not for human or veterinary use.

  • Deuterated internal standard for analytical assays and mass spectrometry.
  • High purity: 99.4%.
  • Molecular formula: C29H32D5N7O5S.
  • Available in small pack sizes (1-100 mg) suitable for method development.
  • Mesylate salt form for improved handling and stability.

Catalog No. 50-002-61997 Supplier Medchemexpress LLC Supplier No. HY142283AS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dosimertinib-d5 mesylate is the deuterated mesylate salt of dosimertinib, a third-generation epidermal growth factor receptor (EGFR) inhibitor. Provided as a research-grade reference standard for preclinical and analytical applications; not for human or veterinary use.

  • Deuterated internal standard for analytical assays and mass spectrometry.
  • High purity: 99.4%.
  • Molecular formula: C29H32D5N7O5S.
  • Available in small pack sizes (1-100 mg) suitable for method development.
  • Mesylate salt form for improved handling and stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.