Learn More
Torpac Inc DOSING APPLICATOR S/S SIZE M
Catalog No. NC2592333
Encompass
If dosing multiple animals, order additional syringes to allow them to dry between animals.
Catalog No. NC2592333 Supplier Torpac Inc Supplier No. XM SYRINGE
If dosing multiple animals, order additional syringes to allow them to dry between animals.