Learn More
METTLER TOLEDO™ Dosing Head Accessory
Catalog No. 01677017 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Quantity:
1 Ea.
10 Pieces
150 Pieces
40 Pieces
500 Pieces
Product Line:
Powder Dosing Head QH002-CNMW
Powder Dosing Head QH008-BNMP
Powder Dosing Head QH010-CNMW
QH012-LNJW Powder Dosing Head
QL003 Liquid Dosing Head
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Line
01-677-017 1 Ea. -
01-676-965 1 Ea. QL003 Liquid Dosing Head
01-677-010 10 Pieces QH012-LNJW Powder Dosing Head
01-677-018 40 Pieces Powder Dosing Head QH010-CNMW
01-677-019 10 Pieces Powder Dosing Head QH010-CNMW
01-677-022 150 Pieces Powder Dosing Head QH010-CNMW
01-677-033 10 Pieces Powder Dosing Head QH008-BNMP
01-677-014 500 Pieces Powder Dosing Head QH002-CNMW
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01-677-017 Supplier METTLER TOLEDO™ Supplier No. 30132792
Add to Cart
Edge
Add to Cart
Dosing accessories efficiently dispense accurate samples for high throughput dosing activities.
  • Automated dispensing in one unit
  • Efficient preparation of accurate samples
  • Built-in RFID chip
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Excellence XPR Automatic Balance
Quantity 1 Ea.
Type Dosing Head Starter Kit