Learn More
BURKLE INC Dosing pump Dosi-Pump, 8.45 oz. (250 ml)stroke, fix. feed.tube
Catalog No. 5000026008
Encompass_Preferred
Dosing pumps are mounted by you in your existing container closure (see the locating bore table). To do this, a suitable hole is drilled in the cap. Then the pump is inserted through the hole and screwed in place from below.
A matching container connection is not included in the scope of delivery.
Catalog No. 50-000-26008 Supplier BURKLE INC Supplier No. 56075250
Dosing pumps are mounted by you in your existing container closure (see the locating bore table). To do this, a suitable hole is drilled in the cap. Then the pump is inserted through the hole and screwed in place from below.
A matching container connection is not included in the scope of delivery.