Learn More
BURKLE INC Dosing pump Dosi-Pump, 1.01 oz. (30 ml) stroke, fix. feed. tube
Catalog No. 5000026012
Encompass_Preferred
Dosing pumps are mounted by you in your existing container closure (see the locating bore table). To do this, a suitable hole is drilled in the cap. Then the pump is inserted through the hole and screwed in place from below.
A matching container connection is not included in the scope of delivery.
Catalog No. 50-000-26012 Supplier BURKLE INC Supplier No. 56073030
Dosing pumps are mounted by you in your existing container closure (see the locating bore table). To do this, a suitable hole is drilled in the cap. Then the pump is inserted through the hole and screwed in place from below.
A matching container connection is not included in the scope of delivery.