Learn More
BURKLE INC Dosing pump Dosi-Pump, 0.14 oz. (4 ml) stroke, imm. depth 9.84 in. (25 cm)
Catalog No. 5000025891
Encompass_Preferred
Dosing pumps are mounted by you in your existing container closure (see the locating bore table). To do this, a suitable hole is drilled in the cap. Then the pump is inserted through the hole and screwed in place from below.
A matching container connection is not included in the scope of delivery.
Catalog No. 50-000-25891 Supplier BURKLE INC Supplier No. 56071004
Dosing pumps are mounted by you in your existing container closure (see the locating bore table). To do this, a suitable hole is drilled in the cap. Then the pump is inserted through the hole and screwed in place from below.
A matching container connection is not included in the scope of delivery.