Learn More
Metrohm USA DOSING UNIT 50MLDOSING UNIT
SDP
Supplier:  Metrohm USA 63032250
Encompass
Dosing unit with integrated data chip with 50 mL glass cylinder and light protection, mountable on reagent bottle with ISO/DIN GL45 thread. FEP tubing connection, anti-diffusion buret tip.
Catalog No. NC0219086