Learn More
Grainger DOT Container Label 4 Label W PK500
Catalog No. 502766070
Encompass_Preferred
The Tripp Lite 16U815 is a 19-inch rack enclosure cabinet with a depth of 15 inches. It has a steel construction with a black powder coat finish and includes a vented steel top panel, vented steel side panels with quick release latches, and a vented steel rear door with a key lock. The cabinet has an adjustable mounting depth from 4 inches to 15 inches and includes mounting hardware. It has an open frame design to allow for side-to-side airflow and is compatible with EIA-310-D rack equipment. The 16U815 provides 15U of internal rack space with a weight capacity of 200 lbs.
Catalog No. 50-276-6070 Supplier Grainger Supplier No. 16U815
The Tripp Lite 16U815 is a 19-inch rack enclosure cabinet with a depth of 15 inches. It has a steel construction with a black powder coat finish and includes a vented steel top panel, vented steel side panels with quick release latches, and a vented steel rear door with a key lock. The cabinet has an adjustable mounting depth from 4 inches to 15 inches and includes mounting hardware. It has an open frame design to allow for side-to-side airflow and is compatible with EIA-310-D rack equipment. The 16U815 provides 15U of internal rack space with a weight capacity of 200 lbs.