Learn More
Grainger DOT Container Label 4 Label W PK500
Catalog No. 502765374
Encompass_Preferred
The DOT Container Label Flammable from Grainger is a 4" x 4" red and white diamond-shaped label that indicates the contents of a container are flammable. It features the word "Flammable" in large bold letters and has space to fill in the proper shipping name, UN/NA number, and flash point. The label is made from vinyl with a permanent adhesive backing and meets DOT and OSHA requirements for labeling flammable materials. It can withstand exposure to many chemicals and solvents. This is an essential safety product to properly identify hazardous materials during storage and transport.
Catalog No. 50-276-5374 Supplier Grainger Supplier No. 16U812
The DOT Container Label Flammable from Grainger is a 4" x 4" red and white diamond-shaped label that indicates the contents of a container are flammable. It features the word "Flammable" in large bold letters and has space to fill in the proper shipping name, UN/NA number, and flash point. The label is made from vinyl with a permanent adhesive backing and meets DOT and OSHA requirements for labeling flammable materials. It can withstand exposure to many chemicals and solvents. This is an essential safety product to properly identify hazardous materials during storage and transport.