Learn More
Labelmaster Inc Flammable Liquid Label, Worded, Paper, Roll Of 500
SDP
Catalog No. NC9212263
Encompass_Preferred
Flammable Liquid DOT Label, Class 3, Printed with Light-Fast Inks for high Durability, 100 x 100mm, Meet the Requirements in 49CFR for Labeling Hazardous Material Shipments, Available in Paper and Vinyl Pressure Sensitive Adhesive, 500/RL
Catalog No. NC9212263 Supplier Labelmaster Inc Supplier No. HML6
Flammable Liquid DOT Label, Class 3, Printed with Light-Fast Inks for high Durability, 100 x 100mm, Meet the Requirements in 49CFR for Labeling Hazardous Material Shipments, Available in Paper and Vinyl Pressure Sensitive Adhesive, 500/RL