Learn More
GA International Cryogenic Color Dots for Tops of 0.5 mL & 1.5 mL Microtubes 0.44" / 11.2mm Circle Yellow From -196°C to +110°C (-321°F to +230°F)
Catalog No. NC0609442
Encompass_Preferred
These small diameter round cryo labels for microtube tops are made with a strong adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen storage. Specially designed to comfortably fit 0.5 ml, 0.6 ml, and 1.5 ml Eppendorf tube caps and microtubes, these circular, waterproof, cryo dot labels have a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. The freezer dot labels are supplied as Hagaki sheets and are available in 15 individual colors as well as packs of 15 assorted colors for color coding and rapid identification.
Catalog No. NC0609442 Supplier GA International Supplier No. LT11YE
Add to Cart
Add to Cart
These small diameter round cryo labels for microtube tops are made with a strong adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen storage. Specially designed to comfortably fit 0.5 ml, 0.6 ml, and 1.5 ml Eppendorf tube caps and microtubes, these circular, waterproof, cryo dot labels have a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. The freezer dot labels are supplied as Hagaki sheets and are available in 15 individual colors as well as packs of 15 assorted colors for color coding and rapid identification.