Learn More
Eurofins Discoverx DOT1L, 50 µg. (EPI018)
Supplier:  Eurofins Discoverx EPI018
Encompass
DOT1L is purified, untagged recombinant DOT1L enzyme. This enzyme contains amino acids 1-420. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5191