Learn More
Medchemexpress LLC DOTA-ADIBO TFA | 1374865-01-6 | 99.1% | 776.76 g/mol | C36H43F3N6O10 | 5 MG
SDP
Catalog No. 5000215674
Encompass_Preferred
DOTA-ADIBO TFA is a DOTA-derived bifunctional chelator supplied as the trifluoroacetic acid salt. It enables biomolecule conjugation via strain-promoted, copper-free azide-alkyne cycloaddition and supports construction of fusion chelator systems for radiometal labeling and PET radiotracer synthesis. The compound is provided as a high-purity solid with DMSO solubility and requires protected, low-temperature storage.
  • DOTA-derived bifunctional chelator for copper-free click conjugation.
  • Facilitates radiometal labeling for PET radiotracer synthesis.
  • High reported purity suitable for research applications.
  • Soluble in DMSO (≈100 mg/mL); may require sonication.
  • Store under nitrogen, protect from light, keep at low temperatures.
  • Provided as a TFA salt to aid handling and stability.
Catalog No. 50-002-15674 Supplier Medchemexpress LLC Supplier No. HY100133A5MG
DOTA-ADIBO TFA is a DOTA-derived bifunctional chelator supplied as the trifluoroacetic acid salt. It enables biomolecule conjugation via strain-promoted, copper-free azide-alkyne cycloaddition and supports construction of fusion chelator systems for radiometal labeling and PET radiotracer synthesis. The compound is provided as a high-purity solid with DMSO solubility and requires protected, low-temperature storage.
  • DOTA-derived bifunctional chelator for copper-free click conjugation.
  • Facilitates radiometal labeling for PET radiotracer synthesis.
  • High reported purity suitable for research applications.
  • Soluble in DMSO (≈100 mg/mL); may require sonication.
  • Store under nitrogen, protect from light, keep at low temperatures.
  • Provided as a TFA salt to aid handling and stability.