Learn More
Bachem 4,7,10-Tricarboxymethyl-1,4,7,10-tetraaza-cyclododecan-1-yl-acetyl-D-Phe-Cys-Tyr-D-Trp-Lys-Thr-Cys-Thr-OH
SDP
Supplier:  Bachem 4046724.0005
Encompass_Preferred
DOTA-(Tyr3)-Octreotate Acetate, 4,7,10-Tricarboxymethyl-1,4,7,10-tetraaza-cyclododecan-1-yl-acetyl-D-Phe-Cys-Tyr-D-Trp-Lys-Thr-Cys-Thr-OH, 177943-89-4, C65H90N14O19S2, Mr 1435.65, 5mg. DOTATATE, DOTA-(Tyr3,Thr8)-Octreotide, DOTA-[Tyr3] Octreotide Acid (Octreotate). DOTA-conjugated somatostatin analog for labeling with radiometals such as gallium-67 or gallium-68, indium-111, and yttrium-90. DOTA-TATE is a useful tool in receptor-mediated tumor imaging and peptide receptor-targeted radionuclide therapy. CAS-Number (net): 177943-88-3.
Catalog No. 50-194-6724