Learn More
Medchemexpress LLC DOTA-cyclo(RGDfK) | 909024-55-1 | 99.5% | 990.07 | 5 MG

Supplier: Medchemexpress LLC HYP107325MG
DOTA-cyclo(RGDfK) is a radionuclide conjugate (RDC) that can bind to a radionuclide. These conjugates are designed to specifically target biomolecules and can be utilized in medical imaging or therapy.
- Solid appearance
- White to off-white color
- Sealed storage, away from moisture
- Powder storage: -80°C for 2 years, -20°C for 1 year
- In solvent storage: -80°C for 6 months, -20°C for 1 month
- Solubility in DMSO: 25 mg/mL (25.25 mM); requires ultrasonic
- Applicable in cancer research and cancer targeted therapy
- Used in radionuclide-drug conjugates (RDCs)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.