Learn More
Medchemexpress LLC Dota-eb-tate | 2151063-66-8 | 95.4% | 2329.63 | 100 UG
SDP
Catalog No. 5000425587
Encompass_Preferred
DOTA-EB-TATE is a peptide drug conjugate (PDC) composed of an SST peptide derivative, DOTA-octreotate, conjugated to an Evans blue analog (EB). It is designed to improve the pharmacokinetics of SSTR2 analogs and reduce PRRT toxicity. This compound can also be used for the synthesis and research of Radionuclide-Drug Conjugates (RDCs).
  • Improves pharmacokinetics of SSTR2 analogs
  • Reduces PRRT toxicity
  • Suitable for synthesis/research of Radionuclide-Drug Conjugates (RDCs)
Catalog No. 50-004-25587 Supplier Medchemexpress LLC Supplier No. HYP10741100UG
DOTA-EB-TATE is a peptide drug conjugate (PDC) composed of an SST peptide derivative, DOTA-octreotate, conjugated to an Evans blue analog (EB). It is designed to improve the pharmacokinetics of SSTR2 analogs and reduce PRRT toxicity. This compound can also be used for the synthesis and research of Radionuclide-Drug Conjugates (RDCs).
  • Improves pharmacokinetics of SSTR2 analogs
  • Reduces PRRT toxicity
  • Suitable for synthesis/research of Radionuclide-Drug Conjugates (RDCs)