Learn More
Medchemexpress LLC DOTA-GA(tBu)4 | 306776-79-4 | MFCD32011828 | 98.7% | 700.90 | C35H64N4O10 | 10 MG
SDP
Catalog No. 5000283956
Encompass_Preferred
DOTA-GA(tBu)4 is a tert-butyl-protected DOTA-GA derivative used as a bifunctional chelator to coordinate radiometals and to prepare DOTA-GA-conjugated compounds for radiolabeling. The tert-butyl protection improves synthetic handling and allows deprotection to the active chelator for downstream labeling workflows.
  • Tert-butyl protection improves stability during chemical synthesis.
  • Designed to facilitate efficient chelation of radiometals for radiolabeling.
  • High purity supports reproducible synthetic and labeling results.
  • Available in small package sizes suitable for research-scale work.
Catalog No. 50-002-83956 Supplier Medchemexpress LLC Supplier No. HYW61426910MG
DOTA-GA(tBu)4 is a tert-butyl-protected DOTA-GA derivative used as a bifunctional chelator to coordinate radiometals and to prepare DOTA-GA-conjugated compounds for radiolabeling. The tert-butyl protection improves synthetic handling and allows deprotection to the active chelator for downstream labeling workflows.
  • Tert-butyl protection improves stability during chemical synthesis.
  • Designed to facilitate efficient chelation of radiometals for radiolabeling.
  • High purity supports reproducible synthetic and labeling results.
  • Available in small package sizes suitable for research-scale work.