Learn More
Medchemexpress LLC DOTA-LM3 TFA | 99.3% | 1664.18 | 5 MG
SDP
Catalog No. 5000398505
Encompass_Preferred
DOTA-LM3 TFA is a somatostatin receptor (SSTR) antagonist. The LM3 component refers to p-Cl-Phe- cyclo(D-Cys-Tyr-D-4-amino-Phe(carbamoyl)-Lys-Thr-Cys)D-Tyr- NH2, which is also a somatostatin antagonist. This product is often isotopically labeled for in vivo tumor tracing, such as 177Lu-DOTA-LM3 TFA and 68Ga-DOTA-LM3 TFA. It can be used for the synthesis and research of Radionuclide-Drug Conjugates (RDCs).
  • Acts as an antagonist for somatostatin receptors
  • Commonly labeled with isotopes like 177Lu or 68Ga for in vivo tumor tracing
  • 68Ga-DOTA-LM3 TFA exhibits favorable biodistribution
  • Shows high tumor uptake and good tumor retention
  • 68Ga-DOTA-LM3 TFA has few safety concerns
  • Useful for research in DOTATOC-negative liver metastases, including pancreatic NET and extensive tumor thrombosis
  • Can be utilized for the synthesis and research of Radionuclide-Drug Conjugates (RDCs)
Catalog No. 50-003-98505 Supplier Medchemexpress LLC Supplier No. HYP5126A5MG
DOTA-LM3 TFA is a somatostatin receptor (SSTR) antagonist. The LM3 component refers to p-Cl-Phe- cyclo(D-Cys-Tyr-D-4-amino-Phe(carbamoyl)-Lys-Thr-Cys)D-Tyr- NH2, which is also a somatostatin antagonist. This product is often isotopically labeled for in vivo tumor tracing, such as 177Lu-DOTA-LM3 TFA and 68Ga-DOTA-LM3 TFA. It can be used for the synthesis and research of Radionuclide-Drug Conjugates (RDCs).
  • Acts as an antagonist for somatostatin receptors
  • Commonly labeled with isotopes like 177Lu or 68Ga for in vivo tumor tracing
  • 68Ga-DOTA-LM3 TFA exhibits favorable biodistribution
  • Shows high tumor uptake and good tumor retention
  • 68Ga-DOTA-LM3 TFA has few safety concerns
  • Useful for research in DOTATOC-negative liver metastases, including pancreatic NET and extensive tumor thrombosis
  • Can be utilized for the synthesis and research of Radionuclide-Drug Conjugates (RDCs)