Learn More
TARGETMOL CHEMICALS INC DOTA-NHS-ester 10MG
SDP
Catalog No. 502878368
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. DOTA-NHS-ester is a linker for affibody molecules with applications in small animal PET, SPECT, and CT. It can be used to label radiotherapeutic agents or imaging probes for tumor detection. Purity ≥98%
Catalog No. 50-287-8368 Supplier TARGETMOL CHEMICALS INC Supplier No. T1784210MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. DOTA-NHS-ester is a linker for affibody molecules with applications in small animal PET, SPECT, and CT. It can be used to label radiotherapeutic agents or imaging probes for tumor detection. Purity ≥98%