Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DOTA-NHS-ester | 170908-81-3 | 501.49 g/mol | C20H31N5O10 | 1 G
SDP

Catalog No. 5000204205
Encompass_Preferred

DOTA-NHS-ester is an amine-reactive DOTA derivative (N-hydroxysuccinimide ester) used to conjugate chelators to peptides, proteins, and other biomolecules for radiolabeling and imaging probe preparation in PET and SPECT studies. It has formula C20H31N5O10 and molecular weight 501.49 g/mol. Manufacturer documents such as a datasheet, SDS, and COA are available.

  • Amine-reactive NHS-ester for conjugation.
  • Suitable for radiolabeling and imaging probe preparation.
  • Chelator compatible with common radiometals.
  • Molecular weight 501.49 g/mol.
  • CAS number 170908-81-3.
  • Supplied in research pack sizes including 1 g.

Catalog No. 50-002-04205 Supplier Medchemexpress LLC Supplier No. HY1288901G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOTA-NHS-ester is an amine-reactive DOTA derivative (N-hydroxysuccinimide ester) used to conjugate chelators to peptides, proteins, and other biomolecules for radiolabeling and imaging probe preparation in PET and SPECT studies. It has formula C20H31N5O10 and molecular weight 501.49 g/mol. Manufacturer documents such as a datasheet, SDS, and COA are available.

  • Amine-reactive NHS-ester for conjugation.
  • Suitable for radiolabeling and imaging probe preparation.
  • Chelator compatible with common radiometals.
  • Molecular weight 501.49 g/mol.
  • CAS number 170908-81-3.
  • Supplied in research pack sizes including 1 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.