Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DOTA-NHS-ester | 170908-81-3 | >97.0% | 501.49 g/mol | C20H31N5O10 | 250 MG
SDP

Catalog No. 5000203554
Encompass_Preferred

DOTA-NHS-ester is an NHS-activated DOTA chelator used to conjugate the DOTA moiety to biomolecules (for example, peptides, proteins, and affibodies) to enable radiolabeling for PET, SPECT, and radiotherapeutic research. It is supplied as a white to off-white solid with high purity and is soluble in DMSO.

  • NHS-activated DOTA chelator for amine conjugation.
  • High purity (>97.0%).
  • Molecular weight 501.49 g/mol; formula C20H31N5O10.
  • Soluble in DMSO (50 mg/mL); ultrasonic assistance recommended.
  • Storage: sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for radiolabeling research (PET/SPECT); not for human use.

Catalog No. 50-002-03554 Supplier Medchemexpress LLC Supplier No. HY128890250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOTA-NHS-ester is an NHS-activated DOTA chelator used to conjugate the DOTA moiety to biomolecules (for example, peptides, proteins, and affibodies) to enable radiolabeling for PET, SPECT, and radiotherapeutic research. It is supplied as a white to off-white solid with high purity and is soluble in DMSO.

  • NHS-activated DOTA chelator for amine conjugation.
  • High purity (>97.0%).
  • Molecular weight 501.49 g/mol; formula C20H31N5O10.
  • Soluble in DMSO (50 mg/mL); ultrasonic assistance recommended.
  • Storage: sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for radiolabeling research (PET/SPECT); not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.