Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DOTA-NHS-ester | 170908-81-3 | MFCD23381190 | ≥97.0% | 501.49 | C20H31N5O10 | 5 G
SDP

Catalog No. 5000202338
Encompass_Preferred

DOTA-NHS-ester is an N-hydroxysuccinimide ester derivative of the DOTA chelator used to conjugate amine-containing molecules for radiolabeling and imaging probe development. It is supplied as a solid and is handled and stored under low-temperature, moisture-free conditions.

  • Used to conjugate DOTA to primary amines for PET and SPECT probe development.
  • High purity suitable for research applications.
  • Soluble in DMSO at 50 mg/mL; protocols provided for aqueous in vivo formulations.
  • Stable as a solid when stored sealed at -20 °C; dissolved samples have recommended cold storage times.
  • Molecular weight 501.49 and CAS number 170908-81-3 for unambiguous identification.

Catalog No. 50-002-02338 Supplier Medchemexpress LLC Supplier No. HY1288905G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOTA-NHS-ester is an N-hydroxysuccinimide ester derivative of the DOTA chelator used to conjugate amine-containing molecules for radiolabeling and imaging probe development. It is supplied as a solid and is handled and stored under low-temperature, moisture-free conditions.

  • Used to conjugate DOTA to primary amines for PET and SPECT probe development.
  • High purity suitable for research applications.
  • Soluble in DMSO at 50 mg/mL; protocols provided for aqueous in vivo formulations.
  • Stable as a solid when stored sealed at -20 °C; dissolved samples have recommended cold storage times.
  • Molecular weight 501.49 and CAS number 170908-81-3 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.