Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DOTA-NHS-ester | 170908-81-3 | MFCD23381190 | >97.0% | 501.49 | C20H31N5O10 | 500 MG
SDP

Catalog No. 5000201925
Encompass_Preferred

DOTA-NHS-ester is a bifunctional chelator containing a DOTA macrocycle and an N-hydroxysuccinimide (NHS) ester. It enables covalent conjugation to amine-containing biomolecules and subsequent radiometal chelation for imaging or radiotherapy research applications.

  • Bifunctional chelator with NHS-ester for amine conjugation.
  • High purity (≥97.0%) solid suitable for research.
  • Molecular weight 501.49 and formula C20H31N5O10.
  • Soluble in DMSO (50 mg/mL) and compatible with common in vivo formulations.
  • Store powder at -20°C; solutions at -80°C (6 months) or -20°C (1 month).
  • Available in a 500 mg pack size.

Catalog No. 50-002-01925 Supplier Medchemexpress LLC Supplier No. HY128890500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOTA-NHS-ester is a bifunctional chelator containing a DOTA macrocycle and an N-hydroxysuccinimide (NHS) ester. It enables covalent conjugation to amine-containing biomolecules and subsequent radiometal chelation for imaging or radiotherapy research applications.

  • Bifunctional chelator with NHS-ester for amine conjugation.
  • High purity (≥97.0%) solid suitable for research.
  • Molecular weight 501.49 and formula C20H31N5O10.
  • Soluble in DMSO (50 mg/mL) and compatible with common in vivo formulations.
  • Store powder at -20°C; solutions at -80°C (6 months) or -20°C (1 month).
  • Available in a 500 mg pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.