Learn More
Medchemexpress LLC DOTA-NHS-ester | 170908-81-3 | MFCD23381190 | >97.0% | 501.49 g/mol | C20H31N5O10 | 50 MG
SDP
Catalog No. 5000203459
Encompass_Preferred
DOTA-NHS-ester is an activated N-hydroxysuccinimide (NHS) ester of the DOTA chelator used to conjugate DOTA to amine-containing molecules for radiolabeling and imaging applications. It is intended for research use and is typically used to prepare peptide or antibody conjugates for small-animal PET, SPECT, and CT studies.
  • Activated NHS-ester for amine conjugation.
  • Used to prepare conjugates for PET, SPECT, and CT imaging.
  • Suitable for labeling radiotherapeutic agents and imaging probes.
  • Molecular weight approximately 501.49 g/mol.
  • Stored sealed and away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-03459 Supplier Medchemexpress LLC Supplier No. HY12889050MG
DOTA-NHS-ester is an activated N-hydroxysuccinimide (NHS) ester of the DOTA chelator used to conjugate DOTA to amine-containing molecules for radiolabeling and imaging applications. It is intended for research use and is typically used to prepare peptide or antibody conjugates for small-animal PET, SPECT, and CT studies.
  • Activated NHS-ester for amine conjugation.
  • Used to prepare conjugates for PET, SPECT, and CT imaging.
  • Suitable for labeling radiotherapeutic agents and imaging probes.
  • Molecular weight approximately 501.49 g/mol.
  • Stored sealed and away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.