Learn More
Medchemexpress LLC DOTA-NHS-ester (TFA) | 00-00-0 | MFCD23381190 | 95.0% | 615.51 g·mol⁻¹ | C22H32F3N5O12 | 5 MG
SDP
Catalog No. 5000264209
Encompass_Preferred
DOTA-NHS-ester TFA is an activated N-hydroxysuccinimide (NHS) ester of the DOTA chelator supplied as the trifluoroacetic acid (TFA) salt. It enables efficient conjugation of the DOTA chelator to amine-containing biomolecules for subsequent radiolabeling and imaging applications such as PET and SPECT.
  • Activated NHS ester for rapid conjugation to primary amines.
  • TFA salt form improves solubility and handling.
  • Facilitates stable metal chelation for radiolabeling.
  • Suitable for labeling peptides, proteins, and affibodies.
  • Available in small-scale research quantities for preclinical work.
Catalog No. 50-002-64209 Supplier Medchemexpress LLC Supplier No. HY128890B5MG
DOTA-NHS-ester TFA is an activated N-hydroxysuccinimide (NHS) ester of the DOTA chelator supplied as the trifluoroacetic acid (TFA) salt. It enables efficient conjugation of the DOTA chelator to amine-containing biomolecules for subsequent radiolabeling and imaging applications such as PET and SPECT.
  • Activated NHS ester for rapid conjugation to primary amines.
  • TFA salt form improves solubility and handling.
  • Facilitates stable metal chelation for radiolabeling.
  • Suitable for labeling peptides, proteins, and affibodies.
  • Available in small-scale research quantities for preclinical work.