Learn More
Cayman Chemical DOTA-NOCacetate 10mg
SDP
Catalog No. 502945514
Encompass_Preferred
A synthetic peptide; has been conjugated to radioligands for use in PET imaging of neuroendocrine tumors and for tissue distribution studies in vivo
Catalog No. 50-294-5514 Supplier Cayman Chemical Supplier No. 4197510 MG
A synthetic peptide; has been conjugated to radioligands for use in PET imaging of neuroendocrine tumors and for tissue distribution studies in vivo