Learn More
CPC Scientific 4,7,10-Tricarboxymethyl-1,4,7,10-tetraaza-cyclododecan-1-yl-acetyl-D-Phe-Cys-Tyr-D-Trp-Lys-Thr-Cys-Thr-OH
(Disulfide bond) 1MG
SDP
Supplier:  CPC Scientific SOMA011A
Encompass
DOTA-conjugated somatostatin analog for labeling with radiometals such as gallium-67 or gallium-68, indiuM-111, and yttrium-90. Potentially useful for receptor-mediated tumor imaging and peptide receptor-targeted radionuclide therapy.SEQUENCE: 4,7,10-Tricarboxymethyl-1,4,7,10-tetraaza-cyclododecan-1-yl-acetyl-D-Phe-Cys-Tyr-D-Trp-Lys-Thr-Cys-Thr-OH(Disulfide bond)ONE-LETTER SEQUENCE: DOTA-fCYwKTCT (Cys & Cys bridge)MOLECULAR FORMULA: C65H90N14O19S2MOLECULAR WEIGHT: 1435.65STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [177943-89-4]SYNONYMS: DOTA-(Tyr)-Octreotate, DOTA-[Tyr3] Octreotide Acid (Octreotate), DOTATATE, DOTA-(Tyr,Thr)-Octreotide, DOTATATERESEARCH AREA: GastrointestinalREFERENCES:W.A.Breeman et al., Eur. J. Nucl. Med. Mol. Imaging, 30, 917 (2003)A.Capello et al., Cancer Biother. Radiopharm., 18, 761 (2003)M.de Jong et al., J. Nucl. Med., 46, 13S (2005)L.Kolby et al., Br. J. Cancer, 93, 1144 (2005)R.Valkema et al., J. Nucl. Med., 4
Catalog No. 50-211-0437