Learn More
Medchemexpress LLC Dota-octreotide tfa | 99.3% | 1519.66 | 1 MG
SDP
Catalog No. 5000425164
Encompass_Preferred
DOTA-Octreotide TFA is composed of the chelator DOTA and Octreotide (HY-P0036). It is utilized for cancer research through its combination with radioactive elements and can be employed for the synthesis and research of Radionuclide-Drug Conjugates (RDCs). Its molecular formula is C67H93F3N14O19S2 with a molecular weight of 1519.66. It appears as a white to off-white solid.
  • Used for cancer research
  • Combines with radioactive elements
  • Can be used for synthesis and research of radionuclide-drug conjugates (RDCs)
  • Soluble in H2O at ≥ 50 mg/mL (32.90 mM)
  • Powder stable for 2 years at -80°C and 1 year at -20°C
  • Solution stable for 6 months at -80°C and 1 month at -20°C
Catalog No. 50-004-25164 Supplier Medchemexpress LLC Supplier No. HYP10273A1MG
DOTA-Octreotide TFA is composed of the chelator DOTA and Octreotide (HY-P0036). It is utilized for cancer research through its combination with radioactive elements and can be employed for the synthesis and research of Radionuclide-Drug Conjugates (RDCs). Its molecular formula is C67H93F3N14O19S2 with a molecular weight of 1519.66. It appears as a white to off-white solid.
  • Used for cancer research
  • Combines with radioactive elements
  • Can be used for synthesis and research of radionuclide-drug conjugates (RDCs)
  • Soluble in H2O at ≥ 50 mg/mL (32.90 mM)
  • Powder stable for 2 years at -80°C and 1 year at -20°C
  • Solution stable for 6 months at -80°C and 1 month at -20°C