Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DOTA-PEG5-amine | 98.0% | 666.76 | 5 MG
SDP

Catalog No. 5000364496
Encompass_Preferred

DOTA-PEG5-amine is a PEG-based PROTAC linker, appearing as a white to off-white solid. It has a molecular weight of 666.76 and a chemical formula of C28H54N6O12. This linker can be utilized in the synthesis of PROTACs, which are small molecules designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.

  • Purity: 98.0%
  • Form: Solid
  • Color: White to off-white
  • Intended for research use only

Catalog No. 50-003-64496 Supplier Medchemexpress LLC Supplier No. 5MGHY140008
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOTA-PEG5-amine is a PEG-based PROTAC linker, appearing as a white to off-white solid. It has a molecular weight of 666.76 and a chemical formula of C28H54N6O12. This linker can be utilized in the synthesis of PROTACs, which are small molecules designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.

  • Purity: 98.0%
  • Form: Solid
  • Color: White to off-white
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.