Learn More
Medchemexpress LLC DOTA-PEG5-amine | 98.0% | 666.76 | 50 MG
SDP
Catalog No. 5000350257
Encompass_Preferred
DOTA-PEG5-amine is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These molecules feature two ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-50257 Supplier Medchemexpress LLC Supplier No. HY14000850MG
DOTA-PEG5-amine is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These molecules feature two ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only