Learn More
Medchemexpress LLC DOTA-PEG5-amine | MFCD31693736 | 98.0% | 666.76 | 1 MG
SDP
Catalog No. 5000349449
Encompass_Preferred
DOTA-PEG5-amine is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs (Proteolysis Targeting Chimeric Molecules). PROTACs are innovative compounds that leverage the cell's ubiquitin-proteasome system to achieve selective degradation of target proteins. These molecules typically comprise two distinct ligands connected by a linker; one ligand is engineered to bind to an E3 ubiquitin ligase, while the other targets the protein intended for degradation.
  • Functions as a PEG-based PROTAC linker.
  • Designed for the synthesis of PROTACs.
  • Enables the selective degradation of target proteins.
  • Utilizes the intracellular ubiquitin-proteasome system.
  • High purity of 98.0%.
Catalog No. 50-003-49449 Supplier Medchemexpress LLC Supplier No. HY1400081MG
DOTA-PEG5-amine is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs (Proteolysis Targeting Chimeric Molecules). PROTACs are innovative compounds that leverage the cell's ubiquitin-proteasome system to achieve selective degradation of target proteins. These molecules typically comprise two distinct ligands connected by a linker; one ligand is engineered to bind to an E3 ubiquitin ligase, while the other targets the protein intended for degradation.
  • Functions as a PEG-based PROTAC linker.
  • Designed for the synthesis of PROTACs.
  • Enables the selective degradation of target proteins.
  • Utilizes the intracellular ubiquitin-proteasome system.
  • High purity of 98.0%.