Learn More
Medchemexpress LLC DOTA-PEG5-azide | 3104986-47-9 | 98.0% | C28H52N8O12 | 100 MG
SDP
Catalog No. 5000353795
Encompass_Preferred
DOTA-PEG5-azide is a PEG-based PROTAC linker designed for synthesizing PROTACs. It functions as a click chemistry reagent, featuring an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Utilized in PROTAC synthesis
Catalog No. 50-003-53795 Supplier Medchemexpress LLC Supplier No. HY140752100MG
DOTA-PEG5-azide is a PEG-based PROTAC linker designed for synthesizing PROTACs. It functions as a click chemistry reagent, featuring an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Utilized in PROTAC synthesis