Learn More
Medchemexpress LLC DOTA-PEG5-azide | 3104986-47-9 | 98.0% | C28H52N8O12 | 25 MG
SDP
Catalog No. 5000352657
Encompass_Preferred
DOTA-PEG5-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Intended for research use only
Catalog No. 50-003-52657 Supplier Medchemexpress LLC Supplier No. 25MGHY140752
DOTA-PEG5-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Intended for research use only