Learn More
Medchemexpress LLC Dota-PEG5-azide | 3104986-47-9 | 98.0% | C28H52N8O12 | 50 MG
SDP
Catalog No. 5000355694
Encompass_Preferred
DOTA-PEG5-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing alkyne groups
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
Catalog No. 50-003-55694 Supplier Medchemexpress LLC Supplier No. HY14075250MG
DOTA-PEG5-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing alkyne groups
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.