Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DOTA-PEG5-C6-DBCO | 97.9% | C49H71N7O14 | 25 MG
SDP

Catalog No. 5000355935
Encompass_Preferred

DOTA-PEG5-C6-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing Azide groups. PROTACs consist of two ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein, utilizing the intracellular ubiquitin-proteasome system for selective protein degradation. This product is for research use only.

  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains a DBCO group for SPAAC with azide groups
  • Molecular weight of 982.13
  • Chemical formula C49H71N7O14
  • Off-white to light yellow solid appearance
  • High purity of 97.9%

Catalog No. 50-003-55935 Supplier Medchemexpress LLC Supplier No. 25MGHY140314
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOTA-PEG5-C6-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing Azide groups. PROTACs consist of two ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein, utilizing the intracellular ubiquitin-proteasome system for selective protein degradation. This product is for research use only.

  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains a DBCO group for SPAAC with azide groups
  • Molecular weight of 982.13
  • Chemical formula C49H71N7O14
  • Off-white to light yellow solid appearance
  • High purity of 97.9%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.