Learn More
Medchemexpress LLC DOTA-(t-butyl)3-PEG5-azide | 00-00-0 | 99.7% | C40H76N8O12 | 10MG
SDP
Catalog No. 5000260175
Encompass_Preferred
DOTA-(t-butyl)3-PEG5-azide is a PEG-based bifunctional chelator linker featuring three tert-butyl-protected DOTA ester groups and a PEG5 spacer terminating in an azide. Supplied as a high-purity research material (99.69%, MW 861.08, formula C40H76N8O12), it is designed for use in bioconjugation, PROTAC linker assembly, and preparation of chelator-conjugated probes.
  • Tri(tert-butyl) DOTA protected as tert-butyl esters for synthetic stability.
  • PEG5 spacer provides flexibility and improves solubility.
  • Azide terminal enables copper-catalyzed and strain-promoted click reactions.
  • High purity suitable for research-grade bioconjugation workflows.
  • Applicable for preparing chelator-conjugated probes and radiolabeling precursors.
Catalog No. 50-002-60175 Supplier Medchemexpress LLC Supplier No. HY14075310MG
Add to Cart
Add to Cart
DOTA-(t-butyl)3-PEG5-azide is a PEG-based bifunctional chelator linker featuring three tert-butyl-protected DOTA ester groups and a PEG5 spacer terminating in an azide. Supplied as a high-purity research material (99.69%, MW 861.08, formula C40H76N8O12), it is designed for use in bioconjugation, PROTAC linker assembly, and preparation of chelator-conjugated probes.
  • Tri(tert-butyl) DOTA protected as tert-butyl esters for synthetic stability.
  • PEG5 spacer provides flexibility and improves solubility.
  • Azide terminal enables copper-catalyzed and strain-promoted click reactions.
  • High purity suitable for research-grade bioconjugation workflows.
  • Applicable for preparing chelator-conjugated probes and radiolabeling precursors.