Learn More
Medchemexpress LLC DOTA-thiol | 865470-67-3 | MFCD11501203 | 98.9% | 463.55 g/mol | C18H33N5O7S | 10 MG
SDP
Catalog No. 5000284554
Encompass_Preferred
DOTA-Thiol is a bifunctional chelator - a macrocyclic DOTA derivative - intended for tumor pre-targeting and for conjugation to peptides and radionuclides. It contains a thiol functional group that enables site-specific attachment and is supplied at high purity for research applications.
  • Bifunctional chelator with a reactive thiol group
  • Enables conjugation of peptides and radionuclides
  • Suitable for tumor pre-targeting and radiolabeling workflows
  • High purity for research-grade use
  • Available in milligram-scale pack sizes for synthesis and labeling
Catalog No. 50-002-84554 Supplier Medchemexpress LLC Supplier No. HYW67839410MG
DOTA-Thiol is a bifunctional chelator - a macrocyclic DOTA derivative - intended for tumor pre-targeting and for conjugation to peptides and radionuclides. It contains a thiol functional group that enables site-specific attachment and is supplied at high purity for research applications.
  • Bifunctional chelator with a reactive thiol group
  • Enables conjugation of peptides and radionuclides
  • Suitable for tumor pre-targeting and radiolabeling workflows
  • High purity for research-grade use
  • Available in milligram-scale pack sizes for synthesis and labeling