Learn More
Medchemexpress LLC DOTA-thiol | 865470-67-3 | MFCD11501203 | 98.9% | 463.55 g/mol | C18H33N5O7S | 5 MG
SDP
Catalog No. 5000284166
Encompass_Preferred
DOTA-thiol is a bifunctional chelator and macrocyclic DOTA derivative used for tumor pre-targeting and radiolabeling research. It provides a thiol functional group for covalent conjugation to peptides, biomolecules, and radionuclides, enabling targeted delivery and imaging studies.
  • Bifunctional macrocyclic chelator suitable for radiolabeling.
  • Thiol functional group enables covalent conjugation to peptides and biomolecules.
  • High purity (98.9%) appropriate for research use.
  • Molecular weight 463.55 g/mol and formula C18H33N5O7S.
  • Available in research-scale quantities such as 5 MG.
Catalog No. 50-002-84166 Supplier Medchemexpress LLC Supplier No. HYW6783945MG
DOTA-thiol is a bifunctional chelator and macrocyclic DOTA derivative used for tumor pre-targeting and radiolabeling research. It provides a thiol functional group for covalent conjugation to peptides, biomolecules, and radionuclides, enabling targeted delivery and imaging studies.
  • Bifunctional macrocyclic chelator suitable for radiolabeling.
  • Thiol functional group enables covalent conjugation to peptides and biomolecules.
  • High purity (98.9%) appropriate for research use.
  • Molecular weight 463.55 g/mol and formula C18H33N5O7S.
  • Available in research-scale quantities such as 5 MG.