Learn More
Medchemexpress LLC DOTA-(Tyr3)-octreotate acetate | 177943-89-4 | MFCD09039058 | 100.0% | 1495.7 g/mol | C65H90N14O19S2 · xC2H4O2 | 1 MG
SDP
Catalog No. 5000244414
Encompass_Preferred
DOTATATE acetate is a DOTA-conjugated somatostatin analogue peptide supplied as the acetate salt for research use. It is formulated for radiolabeling with diagnostic and therapeutic radionuclides and is used in PET imaging research, peptide receptor radionuclide studies, and the synthesis of radionuclide-drug conjugates.
  • High purity suitable for research applications.
  • Solid, white to off-white appearance for easy handling.
  • Optimized for radiolabeling with common PET radionuclides.
  • Supplied in small quantities to support preclinical studies.
  • Stable under low-temperature storage when sealed and protected from moisture.
Catalog No. 50-002-44414 Supplier Medchemexpress LLC Supplier No. HY106244A1MG
DOTATATE acetate is a DOTA-conjugated somatostatin analogue peptide supplied as the acetate salt for research use. It is formulated for radiolabeling with diagnostic and therapeutic radionuclides and is used in PET imaging research, peptide receptor radionuclide studies, and the synthesis of radionuclide-drug conjugates.
  • High purity suitable for research applications.
  • Solid, white to off-white appearance for easy handling.
  • Optimized for radiolabeling with common PET radionuclides.
  • Supplied in small quantities to support preclinical studies.
  • Stable under low-temperature storage when sealed and protected from moisture.