Learn More
CPC Scientific DOTA-DPhe-Cys-Tyr-DTrp-Lys-Thr-Cys-Thr-ol (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific SOMA017B
Encompass
Somatostatin analog DOTA-conjugated for radiolabels such as gallium-67, gallium-68, indium-111, and yttrium-90. This peptide is a useful tool for tumor imaging in radionuclide therapy.SEQUENCE: DOTA-DPhe-Cys-Tyr-DTrp-Lys-Thr-Cys-Thr-ol (trifluoroacetate salt)(Cys2 and 7 bridge)ONE-LETTER SEQUENCE: DOTA-fCYwKTCT-ol (C2&C7 bridge)MOLECULAR FORMULA: C65H90N14O19S2MOLECULAR WEIGHT: 1421.64STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [177943-89-4]SYNONYMS: DOTA-(Tyr3)-Octreotate, DOTATATE, DOTA-(Tyr3,Thr8)-Octreotide, DOTA-[Tyr3] Octreotide Acid (Octreotate)RESEARCH AREA: GastrointestinalREFERENCES:L.Kolby et al., Br. J. Cancer, 93, 1144 (2005)R.Valkema et al., J. Nucl. Med., 46, 83S (2005)M.de Jong et al., J. Nucl. Med., 46, 13S (2005)W.A.Breeman et al., Eur. J. Nucl. Med. Mol. Imaging, 30, 917 (2003)A.Capello et al., Cancer Biother. Radiopharm., 18, 761 (2003)Chinol, M. et al. Semin. Nucl. Med. 32,141(2002)De
Catalog No. 50-211-0443