Learn More
Medchemexpress LLC DOTAP chloride | 132172-61-3 | 99.9% | 698.54 | 25 MG
SDP
Catalog No. 5000439597
Encompass_Preferred
DOTAP chloride is an effective cationic lipid used for transient and stable transfection of DNA (plasmids, bacmids) and modified nucleic acids (antisense oligonucleotides). It facilitates genetic material delivery without the need for a helper lipid.
  • Useful for transient and stable transfection of DNA and modified nucleic acids
  • Cationic lipid
  • Does not require a helper lipid for transfection
Catalog No. 50-004-39597 Supplier Medchemexpress LLC Supplier No. 25MGHY112754AGL
DOTAP chloride is an effective cationic lipid used for transient and stable transfection of DNA (plasmids, bacmids) and modified nucleic acids (antisense oligonucleotides). It facilitates genetic material delivery without the need for a helper lipid.
  • Useful for transient and stable transfection of DNA and modified nucleic acids
  • Cationic lipid
  • Does not require a helper lipid for transfection